missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Optima™ 1 Capillary Column for GC
Brand: Macherey-Nagel™ 726839.50
This item is not returnable.
View return policy
Specifications
0.4 mm | |
0.2 mm | |
G1, G2, G38 | |
340°C to 360°C | |
50 m |
GC Capillary Column | |
Optima™ 1 | |
Separation of Components According to Boiling Points Analysis | |
0.5 μm | |
1 Each |