Learn More
Macherey-Nagel™ Optima™ 1 Capillary Column for GC
Brand:  Macherey-Nagel™ 726050.25
 View more versions of this product
Product Code. 10154461
  • 605.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.25 mm
G1, G2, G38
340°C to 360°C
25 m
GC Capillary Column
Optima™ 1
Separation of Components According to Boiling Points Analysis
0.25 μm
1 Each