Learn More
Macherey-Nagel™ Optima™ 1 Capillary Column for GC
Brand:  Macherey-Nagel™ 726024.20
 View more versions of this product
Product Code. 10421852
  • 545.00 € / Each
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.1 mm
G1, G2, G38
340°C to 360°C
20 m
GC Capillary Column
Optima™ 1
Gas Chromatography (GC)
0.1 μm
1 Each