Learn More
Macherey-Nagel™ Optima™ 1 MS Accent GC Capillary Column
Brand:  Macherey-Nagel™ 725806.60
 View more versions of this product
Product Code. 12422820
  • 1475.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
GC Capillary Column
Optima™ 1 MS Accent
Gas Chromatography (GC)
0.5 μm
1 Each
Ideal for Ion Trap and Quadrupole MS Detectors
0.25 mm
G1, G2, G38
360°C
60 m