Learn More
Macherey-Nagel™ Optima™ 210 GC Capillary Column
375.00 € - 1155.00 €
Specifications
Stationary Phase | Optima™ 210 |
---|---|
USP Type | G1, G2, G38 |
Type | GC Capillary Column |
For Use With (Application) | Gas Chromatography (GC) |
Temperature Range | 260°C to 280°C |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
10680332
|
Macherey-Nagel™
726877.15 |
15 m | 0.32 mm | 0.25 μm |
384.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10596691
|
Macherey-Nagel™
726880.25 |
25 m | 0.32 mm | 0.5 μm |
646.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10082991
|
Macherey-Nagel™
726874.30 |
30 m | 0.25 mm | 0.5 μm |
605.00 €
Each |
In Stock
Log in to see stock. |
|||
10715942
|
Macherey-Nagel™
726871.30 |
30 m | 0.25 mm | 0.25 μm |
605.00 €
Each |
In Stock
Log in to see stock. |
|||
10515571
|
Macherey-Nagel™
726871.25 |
25 m | 0.25 mm | 0.25 μm |
605.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10505571
|
Macherey-Nagel™
726871.15 |
15 m | 0.25 mm | 0.25 μm |
375.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10786502
|
Macherey-Nagel™
726877.30 |
30 m | 0.32 mm | 0.25 μm |
646.00 €
Each |
In Stock
Log in to see stock. |
|||
10516871
|
Macherey-Nagel™
726880.30 |
30 m | 0.32 mm | 0.5 μm |
646.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10216901
|
Macherey-Nagel™
726871.50 |
50 m | 0.25 mm | 0.25 μm |
1005.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10003011
|
Macherey-Nagel™
726874.50 |
50 m | 0.25 mm | 0.5 μm |
1005.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10630722
|
Macherey-Nagel™
726877.50 |
50 m | 0.32 mm | 0.25 μm |
1090.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10618621
|
Macherey-Nagel™
726871.60 |
60 m | 0.25 mm | 0.25 μm |
1050.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10226231
|
Macherey-Nagel™
726877.60 |
60 m | 0.32 mm | 0.25 μm |
1155.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10398351
|
Macherey-Nagel™
726880.50 |
50 m | 0.32 mm | 0.5 μm |
1090.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10368441
|
Macherey-Nagel™
726874.60 |
60 m | 0.25 mm | 0.5 μm |
1050.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10308361
|
Macherey-Nagel™
726880.60 |
60 m | 0.32 mm | 0.5 μm |
1155.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
Specifications
Optima™ 210 | |
GC Capillary Column | |
260°C to 280°C |
G1, G2, G38 | |
Gas Chromatography (GC) | |
1 Each |