Learn More
Macherey-Nagel™ Optima™ 240 GC Capillary Column
Brand:  Macherey-Nagel™ 726091.50
 View more versions of this product
Product Code. 10419551
  • 1090.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.5 mm
0.32 mm
Gas Chromatography (GC), Dioxins
0.25 μm
1 Each
GC Capillary Column
Optima™ 240
260°C to 280°C
50 m