Learn More
Macherey-Nagel™ Optima™ 5 High Performance GC Capillary Column
Brand:  Macherey-Nagel™ 726846.10
 View more versions of this product
Product Code. 10756332
  • 338.00 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.1 mm
G27, G36
340°C to 360°C
10 m
High Performance GC Capillary Column
Optima™ 5
Gas Chromatography (GC)
0.1 μm
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ 5 High Performance GC Capillary Column > Length: 10m; I.D.: 0.1mm; Film Thickness: 0.1um