Learn More
Macherey-Nagel™ Optima™ 5 MS GC Capillary Column
361.00 € - 1335.00 €
Specifications
Stationary Phase | Optima™ 5 MS |
---|---|
USP Type | G27, G36 |
Type | GC Capillary Column |
For Use With (Application) | GC/MS, ECD |
Temperature Range | 320°C to 340°C |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
10556691
|
Macherey-Nagel™
726210.25 |
25 m | 0.2 mm | 0.2 μm |
702.00 €
Each |
In Stock
Log in to see stock. |
|||
10256801
|
Macherey-Nagel™
726210.12 |
12 m | 0.2 mm | 0.2 μm |
361.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10546501
|
Macherey-Nagel™
726215.25 |
25 m | 0.2 mm | 0.35 μm |
702.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10471272
|
Macherey-Nagel™
726212.25 |
25 m | 0.32 mm | 1 μm |
744.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10610342
|
Macherey-Nagel™
726220.30 |
30 m | 0.25 mm | 0.25 μm |
702.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10639381
|
Macherey-Nagel™
726215.12 |
12 m | 0.2 mm | 0.35 μm |
361.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10479161
|
Macherey-Nagel™
726220.15 |
15 m | 0.25 mm | 0.25 μm |
431.00 €
Each |
In Stock
Log in to see stock. |
|||
10506331
|
Macherey-Nagel™
726210.50 |
50 m | 0.2 mm | 0.2 μm |
1145.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10348301
|
Macherey-Nagel™
726211.30 |
30 m | 0.32 mm | 0.25 μm |
744.00 €
Each |
In Stock
Log in to see stock. |
|||
10679951
|
Macherey-Nagel™
726215.50 |
50 m | 0.2 mm | 0.35 μm |
1145.00 €
Each |
In Stock
Log in to see stock. |
|||
10708432
|
Macherey-Nagel™
726220.60 |
60 m | 0.25 mm | 0.25 μm |
1205.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10254092
|
Macherey-Nagel™
726213.30 |
30 m | 0.32 mm | 0.5 μm |
744.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10620342
|
Macherey-Nagel™
726225.30 |
30 m | 0.25 mm | 0.5 μm |
702.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10688991
|
Macherey-Nagel™
726212.50 |
50 m | 0.32 mm | 1 μm |
1245.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10429551
|
Macherey-Nagel™
726226.30 |
30 m | 0.25 mm | 1 μm |
702.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10286221
|
Macherey-Nagel™
726212.60 |
60 m | 0.32 mm | 1 μm |
1335.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10536331
|
Macherey-Nagel™
726225.60 |
60 m | 0.25 mm | 0.5 μm |
1205.00 €
Each |
In Stock
Log in to see stock. |
|||
Specifications
Optima™ 5 MS | |
GC Capillary Column | |
320°C to 340°C |
G27, G36 | |
GC/MS, ECD | |
1 Each |