Learn More
Macherey-Nagel™ Optima™ 5 Performance GC Capillary Column
Brand:  Macherey-Nagel™ 726628.50
 View more versions of this product
Product Code. 10489741
  • 1090.00 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
0.5 mm
0.32 mm
G27
340°C
50 m
High Performance GC Capillary Column
Optima™ 5
Gas Chromatography (GC)
0.35 μm
1 Each