Learn More
Grant Instruments™ Optima Bridge Plate
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
Brand:  Grant Instruments™ T-BR4
 View more versions of this product
Product Code. 12737789
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
  • For use with Grant refrigeration units, heating and cooling circulators
  • Ensures visibility/accessibility of the thermostat whilst optimising bench space
Specifications
Specifications
T Optima Heating Circulator models to R4
Stainless Steel