missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Optima™ TFX200 Series Digital Heated Circulating Bath

Provides outstanding performance for applications requiring accurate temperature control up to a maximum of 200°C. Grant Optima™ TFX200 Series Digital Heated Circulating Bath incorporates Intelligent Control Optimisation™ (ICO) for adaptive intelligent PID temperature control and includes a powerful integral pump.

Brand:  Grant Instruments™ TXF200-ST12

2778.14 EUR valid until 2025-12-31
Use promo code "24306" to get your promotional price.


 View more versions of this product

Product Code. 12787849

  • 2955.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Full colour QVGA TFT screen clearly displaying actual and set temperatures, pump speed and clear status icons
  • TXSEP and TXPEP external probes for monitoring and controllng temperature of remote loads
  • Intuitive screen icons and menus allow fast and accurate setup
  • USB interface allows connection to PC or laptop for programming or data logging
  • Easy to configure automated temperature profiling and control functions
  • TXF200 can store up to 10 programs containing 100 segments
  • Powerful integral pump with multi-stage variable flow rate, maximum flow 23L/min
  • Two point user calibration for optimum accuracy
  • Adjustable over-temperature cut-out Variable high and low temperature alarm settings - configurable to switch a relay
  • Optional Labwise™ PC software for program set-up, data logging and real-time graphing
  • Maximum cooling/heating range (dependent on tank size): ambient +5 to 99°C with plastic (P) tank, or -15° to 200°C with stainless steel (ST) tank
  • Stability ± 0.01°C
Specifications

Specifications

12 L
0°C to 200°C
Stainless Steel Tank
TXF200-ST12
332x 360 x 215 mm
215 mm
120/230 V
50/60 Hz
22 L/min.
USB and RS232 communication interface
530 mbar
±0.01°C
150 mm
300 mm
1840 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.