Learn More
Macherey-Nagel™ Optima™ WAX GC Capillary Column
Brand:  Macherey-Nagel™ 726600.30
 View more versions of this product
Product Code. 10092971
  • 605.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.25 mm
G16
240°C to 250°C
30 m
GC Capillary Column
Optima™ WAX
Gas Chromatography (GC), Aqueous Solutions
0.25 μm
1 Each