Learn More
Macherey-Nagel™ Optima™ WAX GC Capillary Column
Brand:  Macherey-Nagel™ 726549.25
 View more versions of this product
Product Code. 12482840
  • 749.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.8 mm
0.53 mm
G16
240°C to 250°C
25 m
GC Capillary Column
Optima™ WAX
Aqueous Solutions Analysis, Aqueous Solutions
1 μm
1 Each