missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Set of Three Heratrays

Material: Stainless Steel; For Use With: Heracell 150i

Brand:  Thermo Scientific™ 50051913

369.83 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 10006281

  • 413.06 € / Pack of 3
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Material: Stainless Steel; For Use With: Heracell 150i

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
LED
Tray
0.84 cm
Heracell 150i
Stainless Steel
0.33 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.