Learn More
STEMCELL Technologies OptiPrep™
Density gradient medium for the isolation of viruses, organelles, macromolecules, or cells
Brand:  STEMCELL Technologies 07820
Encompass_Preferred
Product Code. 17109821
  • 295.65 € / 250mL
Explore more special offers
Description
Description
  • OptiPrep™ is a density gradient medium recommended for the isolation and purification of DNA or RNA viruses, organelles such as nuclei, mitochondria, endosomes or exosomes, macromolecules, as well as a wide range of cell types
  • OptiPrep™ is a non-ionic iodixanol-based medium with a density of 1.320 ± 0.001 g/mL
  • It has been used clinically in X-ray contrast imaging and has been shown to have lower toxicity compared to ionic gradient compounds such as metrizamide
Specifications
Specifications
250 mL
Mononuclear Cells, Other
Cell Isolation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies OptiPrep™