missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ SmaI, Fisher BioReagents™
107.63 € - 210.00 €
Specifications
Concentration | 10 U/μL |
---|---|
Components | 10X OPTIZYME™ Buffer 4 |
Incubator Temperature | 30°C |
pH | 7.4 |
For Use With (Application) | >90% of pUC19 DNA fragments can be ligated and re-cut after 50-fold over-digestion with SmaI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11944852
|
Fisher BioReagents
BP8011-1 |
1200 U |
210.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11934852
|
Fisher BioReagents
BP8011-5 |
5000 U | N/A | ||||||
Description
5'...C C C^G G G...3'
3'...G G G^C C C...5'
Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
- Incubate at 30°C
- Buffer 1: 50 - 100%
- Buffer 2: 0 - 20%
- Buffer 3: 0 - 20%
- Buffer 4: 100%
- Buffer 5: 0 - 20%
- 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
- Incubation at 37°C results in 50% activity. SmaI has a half-life of 15 min at 37°C.
- Incubation at 25°C results in 100% activity.
- SmaI needs K+ to work for activity.
Dam: Never overlaps - no effect
Dcm: Never overlaps - no effect
CpG: Completely overlaps - blocked
- CCCGGG
Specifications
10 U/μL | |
30°C | |
>90% of pUC19 DNA fragments can be ligated and re-cut after 50-fold over-digestion with SmaI | |
CCC.GGG | |
Liquid |
10X OPTIZYME™ Buffer 4 | |
7.4 | |
10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol | |
SmaI |
Safety and Handling
Product Identifier
- OPTIZYME