Learn More
Hirschmann Instruments™ opus™ Dispenser
Guarantees excellent accuracy during serial dispensing. Hirschmann Instruments™ opus™ Dispenser is for motor-driven dispensing directly from the reagent bottle.
Brand:  Hirschmann Instruments™ 9581020
Product Code. 10752794
  • 3045.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dispenser touch screen module incl. brown UV protection
  • With thread A 45mm, 3 adapters, 1 dispensing discharge unit, 1 power supply
  • With reagent recirculation system
  • Conformity certified
  • Serial and USB interface
Specifications
Specifications
20 mL
TFT touchscreen
100/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann Instruments™ opus™ Dispenser >