missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ ORC Activated Carbon Cartridges

To remove unpleasant tastes and odors of chlorine from water. Fisherbrand™ ORC Cartridges offer a constant and guaranteed level of performance.

Brand:  Fisherbrand™ ORC

Product Code. 11984159

  • 84.15 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Carbon finely divided to capture 100% of chlorine and 98% of organic substances
  • No electrical connection needed
Specifications

Specifications

60 L/hr.
ORC
1 Each
Cartridge
60 mm
For water purification
3 bar
10°C to 40°C
500 mm
1.2 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers