missing translation for 'onlineSavingsMsg'
Learn More

Orcein, for analysis, Thermo Scientific Chemicals

Red dye used in microscopy as a mordant for the staining of flagella, as well as for detecting elastic tissue in sputum.

31.66 € - 86.93 €

Chemical Identifiers

CAS 1400-62-0
Molecular Formula C28H24N2O7
Molecular Weight (g/mol) 500.507
MDL Number MFCD00062310
InChI Key VPEASJIRGSVXBF-UHFFFAOYSA-N
Synonym orcein, pacein, 2,8-bis 2,4-dihydroxy-6-methyl-anilino-1,9-dimethyl-dibenzofuran-3,7-dione, 2,8-bis 2,4-dihydroxy-6-methylanilino-1,9-dimethyldibenzo b,d furan-3,7-dione, unii-655waw52f4, orcein 1g, c.i. natural red 28, ncistruc1_000110, ncistruc2_001941, 2,4-dihydroxy-6-methylphenylamino-1,9-dimethyldibenzo b,d furan-3,7-dione
PubChem CID 5386447
IUPAC Name 2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione
SMILES CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459806
View Documents
Thermo Scientific Alfa Aesar
B20294.03
1 g
31.66 €
1g
Please to purchase this item. Need a web account? Register with us today!
11469806
View Documents
Thermo Scientific Alfa Aesar
B20294.06
5 g
86.93 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is a red dye used in microscopy as a mordant for the staining of flagella, as well as for detecting elastic tissue in sputum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1400-62-0
500.507
VPEASJIRGSVXBF-UHFFFAOYSA-N
5386447
CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O
C28H24N2O7
MFCD00062310
orcein, pacein, 2,8-bis 2,4-dihydroxy-6-methyl-anilino-1,9-dimethyl-dibenzofuran-3,7-dione, 2,8-bis 2,4-dihydroxy-6-methylanilino-1,9-dimethyldibenzo b,d furan-3,7-dione, unii-655waw52f4, orcein 1g, c.i. natural red 28, ncistruc1_000110, ncistruc2_001941, 2,4-dihydroxy-6-methylphenylamino-1,9-dimethyldibenzo b,d furan-3,7-dione
2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione

Specifications

1400-62-0
C28H24N2O7
orcein, pacein, 2,8-bis 2,4-dihydroxy-6-methyl-anilino-1,9-dimethyl-dibenzofuran-3,7-dione, 2,8-bis 2,4-dihydroxy-6-methylanilino-1,9-dimethyldibenzo b,d furan-3,7-dione, unii-655waw52f4, orcein 1g, c.i. natural red 28, ncistruc1_000110, ncistruc2_001941, 2,4-dihydroxy-6-methylphenylamino-1,9-dimethyldibenzo b,d furan-3,7-dione
VPEASJIRGSVXBF-UHFFFAOYSA-N
2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione
5386447
Orcein
Glass bottle
MFCD00062310
Soluble in acetone,alcohol,acetic acid (red color),and dilute aqueous alkali (blue-violet color). Insoluble in water.
CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O
500.507
Powder
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 215-750-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only