missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Orcein, pure

Product Code. p-3734567 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10435091

Brand: Thermo Scientific Chemicals 416570050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

orcein, C28H24N2O7, CAS Number-1400-62-0, Natural Red 28, 25g, 0.98 to 1.18, 06, 885, Black-Purple to Brown, 500.51, Pure, GHS Signal Word: Warning, GHS H Statement: Harmful if swallowed. Causes serious eye irritation. Causes skin irritation. | CAS: 1400-62-0 | C28H24N2O7 | 500.507 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1400-62-0
Molecular Formula C28H24N2O7
Molecular Weight (g/mol) 500.507
MDL Number MFCD00062310
InChI Key VPEASJIRGSVXBF-UHFFFAOYSA-N
Synonym Natural Red 28
PubChem CID 5386447
IUPAC Name 2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione
SMILES CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O

Specifications

Absorption Ratio 0.98 to 1.18
Beilstein 06, 885
Merck Index 15, 6958
Infrared Spectrum Authentic
Lambda Max 575 to 582nm
Formula Weight 500.51
Grade Pure
Packaging Glass bottle
Color Black/Purple to Brown
Quantity 5 g
Physical Form Powder
Chemical Name or Material orcein
Show More Show Less
compliance-icons
Product Identifier
  • Orcein
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.