missing translation for 'onlineSavingsMsg'
Learn More

Orcinol Monohydrate, 99%

Product Code. p-3737126 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10664792

Brand: Thermo Scientific Acros 129550250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6153-39-5 | C7H8O2·H2O | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-39-5
Molecular Formula C7H8O2·H2O
Molecular Weight (g/mol) 142.15
MDL Number MFCD00149092
InChI Key NBKPNAMTHBIMLA-UHFFFAOYSA-N
Synonym 5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate
PubChem CID 3083941
IUPAC Name 5-methylbenzene-1,3-diol;hydrate
SMILES CC1=CC(=CC(=C1)O)O.O

Specifications

Melting Point 56.0°C to 61.0°C
Color White
Boiling Point 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 06, 882
Merck Index 14, 6864
Solubility Information (5% in water) Clear colorless to light brown
Formula Weight 142.15
Percent Purity 99%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material Orcinol monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Orcinol monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.