missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Org 24598, >98%, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective inhibitor of the glial glycine transporter GlyT1b (IC50 = 6.9 nM). Displays negligible activity at GlyT-2, adrenoreceptors, dopamine, 5HT receptors and noradrenaline, dopamine, 5HT and GABA transporters (pIC50 < 4). Active in vivo.
Specifications
Specifications
Linear Formula | C19H20F3NO3 |
Quantity | 50 mg |
Solubility Information | Soluble to 10mM in water with gentle warming and to 100mM in DMSO |
Formula Weight | 367.36 |
Chemical Name or Material | N-Methyl-N-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-glycine |
Grade | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction