Learn More
3M™ Organic Vapor Monitors
Lightweight, self-contained monitors record time-weighted average (TWA) vapor concentrations in factories and industrial, histology, cytology, and clinical labs
113.00 € - 1565.70 €
Specifications
For Use With (Application) Periodic Sampling and Screening of hazardous areas and jobs, indicating trouble spots, Checking ventilation, and keeping accurate employee health records
Certifications/Compliance OSHA
Detectable Analytes Organic Vapor
View More Specs
Products 4
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15730018
View Documents
3M™
7000144896
Organic Vapor Monitor This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15720038
View Documents
3M™
7100011298
Organic Vapor Monitor with Prepaid Analysis This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10054324
View Documents
3M™
7000001878
Organic Vapor Monitor only This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15779767
View Documents
3M™
7000051865
Organic Vapor Monitor with Backup and Analysis This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Provide low per-test cost. Short-term and workshift sampling can be conducted easily so you can quantify vapor exposure by employee, job category, or area.
  •  
  • Usage
  •  
    • To use, simply remove a monitor from its package and clip it to clothing near the breathing zone
    • For monitoring a specific work area, place the monitor in a location where airflow is unrestricted
    • After monitoring, seal the badge with the supplied cap; replace it in the package; record the time, date, and vapors to be analyzed; and send it to the lab for analysis†
    •  
    • Applications
    • Periodic sampling and screening of hazardous areas and jobs; indicating trouble spots; checking ventilation; and keeping accurate employee health records.
    • Monitor Types
      • Monitors 3500 and 3510 are designed to provide precise TWA concentration measurements for an eight-hour period. Monitors 3500 and 3520 allow you to collect data for analysis by an independent laboratory. Monitors 3510 and 3530 include prepaid analysis for up to three compounds per badge
      • Monitors 3520 and 3530 are designed with a special backup section for sampling environments that include compounds like acetone, methylene chloride, and isopropyl alcohol
      • Acetone, methyl ethyl ketone, and vinyl acetate may show a decreased recovery when sampled in high relative humidity. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
      • 3M Catalog# 3500, 3510, 3520, 3530
      Misuse may result in sickness or death.
      Order Info
      Data is available from 3M on sampling rate and measuring range for more than 135 vapors that these monitors can sample. Contact your Fisher Scientific Customer Service Representative at 1-800-772-6733 for a complete list of organic compounds sampled.
      Compliance
      Monitors meet or exceed OSHA accuracy requirements of ±25% for many workplace contaminants.
    Specifications
    Specifications
    Periodic Sampling and Screening of hazardous areas and jobs, indicating trouble spots, Checking ventilation, and keeping accurate employee health records
    OSHA
    Organic Vapor
    Videos
    Safety and Handling
    Safety and Handling
    missing translation for 'shelfLife' : 18 Months