missing translation for 'onlineSavingsMsg'
Learn More

Organometallic Oil Standard, 21-Element, 500μg/g, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

257.00 € - 397.00 €

Specifications

Chemical Name or Material 21 Element Oil Standard
Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11460349
View Documents
SPEX Certiprep
S21-500Z
50 g Triple-leached LDPE bottle
257.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
11450349
View Documents
SPEX Certiprep
S21-500Y
100 g Triple-leached LDPE bottle
397.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag500μg/g
Al500μg/g
B500μg/g
Ba500μg/g
Ca500μg/g
Cd500μg/g
Cr500μg/g
Cu500μg/g
Fe500μg/g
Mg500μg/g
Mn500μg/g
Mo500μg/g
Na500μg/g
Ni500μg/g
P500μg/g
Pb500μg/g
Si500μg/g
Sn500μg/g
Ti500μg/g
V500μg/g
Zn500μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

21 Element Oil Standard
With Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Each component at 100μg/g (S21-100), 300μg/g (S21-300), 500μg/g (S21-500), or 900μg/g (S21-900) in Base Oil 75:
Aluminum
Barium
Boron
Cadmium
Calcium
Chromium
Copper
Iron
Lead
Magnesium
Manganese
Molybdenum
Nickel
Phosphorous
Silicon
Silver
Sodium
Tin
Titanium
Vanadium
Zinc
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Oil
Base Oil 75
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only