missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer

Control and optimize boiler ammonia levels with continuous, direct and precise ammonia measurements from the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.

Brand:  Thermo Scientific™ 2110XPEN


 View more versions of this product

Product Code. 17151801

  • 16570.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Reduce operational costs while producing the highest purity steam with the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer. This analyzer provides continuous, direct and precise measurements for the control and optimization of boiler ammonia levels.

The Orion 2110XP indicates the most sensitive changes in ammonia concentration for condensate and feedwater optimization and control.

  • Accurate detection allows precise ammonia control with confidence — maintain proper chemical feed levels without the guess work or excessive operating cost
  • Stable and drift-free measurements with minimal maintenance and eliminates the need for frequent calibrations, saving you time and money
  • Repeatable and verifiable ammonia measurements using superior Orion™ ammonia electrode technology for the highest level of performance available
  • Designed for ammonia specificity, the unique chemistry provides unsurpassed results across a wide range without the pitfalls of pH and conductivity “bottom out effects”
  • Easy to operate and calibration — the Orion™ 2110XP walks you through the step-by-step calibration process, ensuring a successful calibration the first time and every time
  • Ammonia detection range of 0 to 10ppm for the most direct, accurate and precise results available
  • Continuous online measurement ensures real-time information for optimum contorl of the ammonia feed without excessive operating costs
  • Selective and sensitive for ammonia, our system surpasses the performance of other systems that have the limitations of pH and conductivity to track real-time boiler ammonia levels

2110XP Ammonia Analyzer Ordering Information

  • Annual Consumables Kit, Cat. No. 211050XP
  • Reagent, Cat. No. 181030
  • Diffusion Tubing, Cat. No. 150060
  • Calibration Kit, Cat. No. 2110XPCAL
  • Calibration Standards, Cat. No. 181040
  • Ammonia Electrode, Cat. No. 100047
  • Reference Electrode, Cat. No. 210056
  • Reference Electrode Filling Solution, Cat. No. 181073
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±15% or 30 ppb, whichever is greater
±0.5°C
2, Galvanically Isolated
Valox 364
1
2
5°C to 35°C
Custom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
40mL/min.
8 to 100psig
Membrane
0 to 10ppm
0 to 20 mA; 4 to 20 mA
3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
0.1°C
1/4 in. NPTF tube fitting
5 to 45°C
11 in.
17 in.
26 in.
85 to 132 VAC; 170 to 264 VAC
0 to 60 Hz
Optional ph/ORP or Conductivity
± (0.5 mV + 0.1%)
3
Double Known Addition
CE, CSA, UL, FCC Class A limits
Modbus with Additional Module
2110XP Ammonia Analyzer with Protective Enclosure
Ammonia Analyzer
Waterproof; IP66; NEMA 4X
2110XP ammonia analyzer with Ammonia electrode, reference electrode (100056), ATC probe (2100TP),ammonia calibration standards kit, CD user guide and options kit
85-132; 170-264 VAC
5 to 95%, non-condensing
Wall mount
±1999.9m
0.1mV
95% within 2 minutes of injecting a standard solution
1
11 x 17 x 26 in. (27 x 45 x 65cm)
27 cm
45 cm
65 cm
5A
Ammonia
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.