Learn More
Thermo Scientific™ Orion™ 8135BN ROSS™ Combination Flat Surface pH Electrode
Get stable readings to 0.01 pH in less than 30 seconds with this combination flat surface pH electrode.
Marca: Thermo Scientific™ 8135BN
296.65 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Descrizione
Get stable readings to 0.01 pH in less than 30 seconds with the Thermo Scientific™ Orion™ 8135BN ROSS™ Combination Flat Surface pH Electrode. This instrument provides results three to five times more precise than those obtained with conventional electrodes.
The 8135BN ROSS Combination Flat Surface pH Electrode measures pH of soft, moist surfaces — both solid and semi-solid —such as agar gel plates, meats, bread dough and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50°C or more.
- Material: Epoxy body
- pH range: 0 to 14
- Temperature range: 0° to 100°C
- Internal reference: ROSS
- Junction: Glass fiber
- Dimensions: 120 × 12mm
- pH precision: 0.01
- Fill solution: Orion 810007
- Cap diameter: 16mm
- Small samples are available in micro sample dish
- Flat surface tip
Includes: One 60 ml bottle of ROSS fill Solution, 810007 and spout cap
Specifica
With BNC connector | |
Glass Fiber | |
0 to 14 | |
Combination Flat Surface pH Electrode, BNC connector | |
3M KCl | |
ROSS | |
Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode. | |
39 in. | |
0.47 in. | |
16 mm |
BNC | |
Epoxy Body | |
0°C to 100°C | |
<0.002pH/day | |
Standard pH Meter, BNC connector | |
0.01 | |
32°F to 212°F | |
1 m | |
12 mm |