missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ 927005MD Epoxy ATC Probe

Get fast, accurate measurements in lab or field with this ATC Probe, which automatically corrects pH measurements for variation in electrode slope.

Brand:  Thermo Scientific™ 917005

 View more versions of this product

Product Code. 10160921

  • 412.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Get fast, accurate measurements in the lab or field with the Thermo Scientific™ Orion™ 927005MD Epoxy ATC Probe. This instrument automatically corrects pH measurements for variation in electrode slope due to temperature change.

Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATC Probe with 8 Pin DIN connector
Orion Star Series A Meters
0°C to 80°C
0 to 14
6 mm
120 mm
±1°C
Epoxy
ATC Probe, Epoxy
0.2 in.
4.7 in.
32°F to 176°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.