Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Ensure your turbidity meter has the accessories it needs, including turbidity vials, primary turbidity standards, Formazin turbidity standards, and more.
Brand:  Thermo Scientific™ AC2T24
 View more versions of this product
Product Code. 11722178
  • 180.90 € / Pack of 12
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure your Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter has the accessories it needs with our selection of Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories that include turbidity vials, primary turbidity standards, Formazin turbidity standards, and more. Our Orion AQUAfast AQ4500 turbidity meter has advanced features, including dual light sources and drinking water / wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols; use the White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.
Log data into internal memory, or order AQ4CBL cable to send data to printer or download to a computer for easy time and date stamped results storage.
  • U.S. EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg. 38351-38356
  • Dual source to comply with EPA 180.1 and ISO 7027
  • Nephelometric and ratiometric measurements with autoranging
  • IP67 waterproof portable turbidity meter
  • Readings in the range of 0 to 1000 NTU with a choice of units: NT, FTU, FNU, ASBC or EBC
Includes:
  • AQ4500 meter
  • Set of 5 USEPA approved primary calibration standards
  • Seven glass vials
  • User guide
  • Carrying case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Turbidity Vial
AQ4500 Turbidimeter
Replacement Turbidity Vials
US EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg 38351-38356.EPA 180.1 and ISO 7029
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.