Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Ensure your turbidity meter has the accessories it needs, including turbidity vials, primary turbidity standards, Formazin turbidity standards, and more.
29.25 € - 342.71 €
Products 5
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11757717
View Documents
Thermo Scientific™
AC45FZ
Formazin turbidity standard, 4000NTU, 475mL
88.64 €
475mL
In Stock
to see stock.
Add to basket
10494362
View Documents
Thermo Scientific™
AC45ST
Primary Standards for AQ4500, SDVB for 0, 1.0, 10, 100, 1000NTU
342.71 €
Each
In Stock
to see stock.
Add to basket
11767717
View Documents
Thermo Scientific™
AC45S1
Silicone Oil and Cloth Kit
29.25 €
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
11722178
View Documents
Thermo Scientific™
AC2T24
Replacement Turbidity Vials
180.90 €
Pack of 12
In Stock
to see stock.
Add to basket
10672853
View Documents
Thermo Scientific™
AQ4CBL
RS232 cable for method uploads and data downloads in AQ4000/AQ4500
212.06 €
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
Description
Description
Ensure your Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter has the accessories it needs with our selection of Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories that include turbidity vials, primary turbidity standards, Formazin turbidity standards, and more. Our Orion AQUAfast AQ4500 turbidity meter has advanced features, including dual light sources and drinking water / wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols; use the White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.
Accessories for the Orion AQUAfast turbidity meter include primary standards, Formazin turbidity standard, silicon oil and cloth kit, and replacement turbidity vials.
Warranty: Out of box
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Formazin Standard, 4000 NTU
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see supplemental first aid instructions on this label).
Supplemental information
  • MIXTURE LIST-Contains : Hexamethylenetetramine
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.