Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Brand:  Thermo Scientific™ AC2065
 View more versions of this product
Product Code. 11722228
  • 40.50 € / Pack of 50
Explore more special offers
Description
Description
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zinc
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
Set of 2: 50 Copper/Zinc LR Tablets, 50 EDTA Table
Colorimetric water and wastewater testing equipment
0.02 to 1 mg/L Zn
50 Tests
Tablet
AQUAfast zinc reagent tablets, 50 tests
Zincon
AQUAfast 24mm round vials (Cat. No. AC2V24)
Wastewater: Standard Methods 3500-Zn B
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits > Test: Copper/Zinc
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.