missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits

Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.

Brand:  Thermo Scientific™ AC2007


 View more versions of this product

Product Code. 11732218

  • 34.87 € / Pack of 50
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitrate
Orion AQUAfast AQ4000 Colorimeter
50 x Nitrite HR No. 1 Tablets, 50 x Nitrite HR No. 2 Tablets
Colorimetric water and wastewater testing equipment
1 to 40 mg/L
50 Tests
Tablet
AQUAfast nitrate reagent tablets, 50 tests
Zinc Reduction
AQUAfast 24mm round vials (Cat. No. AC2V24)
Drinking Water: ASTM D3867-90 (B) / Standard Methods 4500-NO3-E; Wastewater: ASTM D3867-04 (B) / Standard Methods 4500-NO3-E
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • AQUAFAST II NITRATE TAB.

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ammonium chloride, zinc powder

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.