Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Brand: Thermo Scientific™ AC2061
Description
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
- For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
- No. of tests: 50 to 100
Specifications
Silica | |
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast | |
100 Silica Phosphate Removal Tablets | |
Colorimetric water and wastewater testing equipment | |
0.05 to 4 mg/L SiO2 | |
100 Tablets |
Tablet | |
AQUAfast silica phosphate removal reagent, 100 tablets | |
Phosphate Removal | |
AQUAfast 24mm round vials (Cat. No. AC2V24) | |
Drinking Water: ASTM D859-05,-10 / Standard Methods 4500-SiO2 D; Wastewater: ASTM D859-05 |
Safety and Handling
- silica PR
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : tartaric acid, boricacid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.