missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits

Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.

Brand:  Thermo Scientific™ AC2017


 View more versions of this product

Product Code. 11772218

  • 28.34 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chloride
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
Set of 2: 50 Chloride T1 Tablets, 50 Chloride T2 T
Colorimetric water and wastewater testing equipment
0.5 to 25 mg/L Cl-
Tablet
AQUAfast chloride reagent tablets, 50 tests
Silver Nitrate/Turbidity
AQUAfast 24mm round vials (Cat. No. AC2V24)
50 Tests
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CHLORIDE T1

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sodium bisulfate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.