Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Brand: Thermo Scientific™ AC2027
Description
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
- For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
- No. of tests: 50 to 100
Specifications
Aluminum | |
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast | |
Set of 2: 50 Aluminum No 1 Tablets, 50 Aluminum No | |
Colorimetric water and wastewater testing equipment | |
0.01 to 0.3 mg/L Al | |
50 Tests |
Tablet | |
AQUAfast aluminum reagent tablets, 50 tests | |
Eriochrome Cyanine R | |
AQUAfast 24mm round vials (Cat. No. AC2V24) | |
Wastewater: Standard Methods 3500-Al B-2001 |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
- Skin sensitiser Category 1
Hazard Statement
- H228-Flammable solid.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : methenamine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.