Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Brand:  Thermo Scientific™ AC2035
 View more versions of this product
Product Code. 15109192
  • 29.15 € / Pack of 100
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
For use with Orion AQUAfast™ AQ4000, Orion AQUAfast™ AQ3070, Orion AQUAfast™ AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters.
  • Number of tests: 50 to 100
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bromine
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter
0.1 to 13.5 mg/L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • AQUAFAST II REAGENT -BROMINE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetone
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.