Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Brand:  Thermo Scientific™ AC3072
 View more versions of this product
Product Code. 15149192
  • 83.21 € / Pack of 100
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and other colorimeters
  • No. of tests: 50 to 100
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chlorine, Sodium Chlorate
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter
AQUAfast chlorine (total) HR & sodium chlorate reagent tablets, 100 tests
5 to 200 mg/L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.