Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifications
Specifications
| Type | Aluminum |
| Test Reagent Formats | Ampoule |
| For Use With (Equipment) | Orion AQUAfast AQ4000 Colorimeter |
| Description | AQUAfast aluminum Auto-ID ampoules, 30 pack |
| Includes | 30 Auto-ID Ampoules |
| Method | Eriochrome Cyanine R |
| For Use With (Application) | Colorimetric water and wastewater testing equipment |
| Recommended Accessories | AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) |
| Range | 0.04 to 0.25 mg/L Al |
| EPA Approved Methods | Wastewater: Standard Methods 3500-Al B-2001 |
| Show More |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.