Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Brand:  Thermo Scientific™ AC4007
 View more versions of this product
Product Code. 11722198
  • 135.68 € / 30 tests
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nitrate
Orion AQUAfast AQ4000 Colorimeter
30 Auto-ID Ampoules
Colorimetric water and wastewater testing equipment
5 to 50 mg/L NO3
30 Ampoules
Ampoule
AQUAfast nitrate HR Auto-ID ampoules, 30 pack
Cadmium Reduction
AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Drinking Water: ASTM D3867-90 (B) / Standard Methods 4500-NO3- E; Wastewater: ASTM D3867-04 (B) / Standard Methods 4500-NO3- E
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Nitrate 3 (as NO3)-HR
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
Supplemental information
  • MIXTURE LIST-Contains : ethelene glycol, isopropyl alcohol, sulphanilic acid
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.