missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents

Product Code. 11772198
Click to view available options
Type:
Aluminum
Ammonia
Chloride
Chlorine
Chlorine Dioxide
Copper
Iron
Manganese
Nitrate
Nitrite
Ozone
Phosphate
Silica
Sulfate
Sulfide
Zinc
Range:
0.04 to 0.25 mg/L Al
0.08 to 1 mg/L N
0.2 to 1.5 mg/L N
0.2 to 3 mg/L N
0.2 to 3 mg/L S
0.2 to 5 mg/L O3
0.2 to 6 mg/L Fe
0.3 to 3 mg/L Zn
0.3 to 8 mg/L PO4
0.4 to 5 mg/L Cl2
0.5 to 10 mg/L SiO2
0.5 to 12 mg/L Cu
0.5 to 7 mg/L N
0.8 to 11 mg/L ClO2
1.0 to 7.5 mg/L N
1.5 to 14 mg/L N
10 to 100 mg/L SO4
2 to 30 mg/L Mn
2.5 to 40 mg/L Cl-
5 to 50 mg/L NO3
Method:
Bariumsulfate-Turbidity
Bathocuproine
Cadmium Reduction
DPD
Diazotization (Azo)
Direct Nesslerization
Eriochrome Cyanine R
Ferric Thiocyanide
Heteropoly Blue
Methylene Blue
Periodate
Phenanthroline
Salicylate
Stannous Chloride
Zincon
Unit Size:
30 tests
Pack of 30
This item is not returnable. View return policy

Product Code. 11772198

Brand: Thermo Scientific™ AC4012

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.

Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.

  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
TRUSTED_SUSTAINABILITY

Tekniske data

Type Ammonia
Test Reagent Formats Ampoule
For Use With (Equipment) Orion AQUAfast AQ4000 Colorimeter
Description AQUAfast ammonia LR Auto-ID ampoules, 30 pack
Includes 30 Auto-ID Ampoules
Method Direct Nesslerization
For Use With (Application) Colorimetric water and wastewater testing equipment
Recommended Accessories AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Range 0.5 to 7 mg/L N
EPA Approved Methods Wastewater: ASTM D1426-08 (A)
Quantity 30 Ampoules
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Orion AQUAfast IV Chemistry Reagents
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Faresætning
  • H290-May be corrosive to metals.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Sikkerhedssætning
  • P201-Obtain special instructions before use.
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see supplementary firstaid instructions on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P361-Take off immediately all contaminated clothing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P390-Absorb spillage to prevent material damage.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplerende oplysninger
  • MIXTURE LIST-Contains : sodium hydroxide, tetrasodium diborate,mercury iodide, potasium iodide

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.