Learn More
Thermo Scientific™ Orion™ AquaMate Preprogrammed Methods on USB Memory Stick
Use the Thermo Scientific™ Orion™ AquaMate Preprogrammed Methods on USB Memory Stick with over 260 preprogrammed methods on the Orion™ AquaMate spectrophotometers
Brand:  Thermo Scientific™ AQ0USB
Product Code. 14805071
  • 67.30 € / Each
Explore more special offers
Description
Description
Use the Thermo Scientific™ Orion™ AquaMate Preprogrammed Methods on USB Memory Stick with over 260 preprogrammed methods for Orion™, CHEMetrics and Merck reagent chemistries on the Orion™ AquaMate 8000 UV-Vis and Orion™ AquaMate 7000 Vis spectrophotometers. Preprogrammed methods can be added, removed or modified on the Orion™ AquaMate spectrophotometers at any time.
  • Compatible with the AquaMate 8000 UV-Vis and AquaMate 7000 Vis Spectrophotometers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USB Memory Stick
Orion AquaMate spectrophotometers
Over 260 preprogrammed methods compatible with several leading brands of reagents