Learn More
Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Optimized for your water analysis needs AquaMate 7100 Vis and AquaMate 8100 UV-Vis Spectrophotometers provide fast and reliable results
4693.35 € - 8150.55 €
Specifications
Certifications/Compliance CE / TUV
Compartments Accessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height Removable, washable sample compartment liner with magnetic placement and hold-down
Connections Single USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Depth (Metric) 35.5 cm
Detector Type Dual Silicon Photodiodes
View More Specs
Includes: Test Tube Holder for vials and test tube from 12 to 25 mm OD, AquaMate Water Analysis Methods Software, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362)
Products 3
Product Code Brand Description Spectral Bandwidth Price Quantity & Availability  
Product Code Brand Description Spectral Bandwidth Price Quantity & Availability  
17143841
View Documents
Thermo Scientific™
AQ7100
AquaMate 7100 Vis Spectrophotometer with North America, European and UK Power cord 5 nm
4693.35 €
Each
16439835
View Documents
Thermo Scientific™
AQ8100
AquaMate 8100 UV-Vis Spectrophotometer with North America, European and UK Power cord 2 nm
8150.55 €
Each
17314773
View Documents
Thermo Scientific™
AQ8100APAC
AquaMate 8100 Vis Spectrophotometer with China, Indian and Australia / NZ Power cord 2 nm
8150.55 €
Each
Description
Show More
Specifications
Show More
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis Spectrophotometers