Learn More
Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler Accessories
Maintain automated sample analysis with accessories for our legacy autosampler.
394.00 € - 473.00 €
Specifications
Type Titrator Accessories
View More Specs
Products 2
Product Code Brand Container Size Price Quantity & Availability  
Product Code Brand Container Size Price Quantity & Availability  
11762228
View Documents
Thermo Scientific™
ASB050
50 mL beakers This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10150971
View Documents
Thermo Scientific™
ASB150
150 mL beakers This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Maintain automated sample analysis with accessories for the legacy Thermo Scientific™ Orion™ AutoTration 500 Autosampler. With a choice of 48- and 28-position racks, this products offers flexibility with interchangeable racks to accommodate sample size and volume.
  • Interchangeable racks to accommodate sample size and volume
  • Choice of 48- and 28-position racks
  • Beaker sizes of 45-, 50- and 150-mL
  • Replacement electrode holders for each model
Specifications
Specifications
Titrator Accessories
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.