missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Bromide Electrodes

Measure bromide ions in aqueous solutions quickly, simply, accurately and economically.

815.06 € - 950.73 €

Specifications

Junction Free-flowing Sure-Flow junction assures stable, drift-free potentials
For Use With (Equipment) ISE Meters
View More Specs

Products 2
Product Code Brand Connector Types Price Quantity & Availability  
Product Code Brand Connector Types Price Quantity & Availability  
10212591
View Documents
Thermo Scientific™
9435BN
BNC
950.73 €
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
10314721
View Documents
Thermo Scientific™
9635BNWP
Waterproof BNC
815.06 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Measure bromide ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Bromide Electrodes.

Features
9635BNWP is a combination electrode with  a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
 
Solutions required
Standard
943506     0.1 M bromide standard, 475 mL bottle
 
ISA
940011     ISA for bromide measurements, 475 mL bottle
 
Electrode fill solution
900063     Optimum Results D electrode filling solution, 5 x 60 mL bottles
  
Items included with
900063     Optimum Results D electrode filling solution, 60 mL 1 bottle
948201     Polishing strip. Qty 1
 
Warranty period
12 months from date of purchase
Specifications

Specifications

Free-flowing Sure-Flow junction assures stable, drift-free potentials
ISE Meters
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.