Learn More
Thermo Scientific™ Orion™ Conductivity Standards and Solutions
Descripción
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.
Conductivity Standards
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
- Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
Especificaciones
Especificaciones
| Description | Conductivity Standard |
| Packaging | Bottles |
| Traceability to NIST | Yes |
| Conductivity | 111.9 mS/cm |
| Color | Clear |
| pH | 10 to 12 |
| Boiling Point | 100°C |
| Solubility | Soluble |
| Solution Type | Standard Solution |
| Type | Conductivity Standard |
| Mostrar más |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.