missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Conductivity Standards and Solutions

Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.

49.30 € - 142.71 €

Specifications

Traceability to NIST Yes
Type Conductivity Standard
Boiling Point 100°C
Solubility Soluble
View More Specs

Products 9
Product Code Brand Packaging Concentration Conductivity Price Quantity & Availability  
Product Code Brand Packaging Concentration Conductivity Price Quantity & Availability  
10616453
View Documents
Thermo Scientific™
011001
Bottle
80.80 €
Each
Please to purchase this item. Need a web account? Register with us today!
10364141
View Documents
Thermo Scientific™
011008
Bottle 47 ppm as NaCl 100 μS/cm
128.64 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
11906079
View Documents
Thermo Scientific™
011007
Bottle 692 ppm as NaCl 1413 μS/cm
133.67 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
 
10075463
View Documents
Thermo Scientific™
011005
Bottles 692 ppm as NaCl 111.9 mS/cm
142.71 €
Pack of 5
In Stock
to see stock.
Add to basket
 
11916079
View Documents
Thermo Scientific™
011006
Bottle 7230 ppm as NaCl 12.9 mS/cm
128.64 €
Pack of 5
In Stock
to see stock.
Add to basket
 
11747717
View Documents
Thermo Scientific™
990106
Bottle 0.1M KCl 12.9 mS
97.79 €
475mL
Please to purchase this item. Need a web account? Register with us today!
11717817
View Documents
Thermo Scientific™
01100910
Pouch 147 μS/cm
50.15 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11707817
View Documents
Thermo Scientific™
01100710
Pouch 692 ppm as NaCl 1413 μS/cm
49.30 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11797807
View Documents
Thermo Scientific™
01100610
Pouch 7230 ppm as NaCl 12.9 mS/cm
50.15 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.

Conductivity Standards

  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
  • For use with Thermo Scientific™ Orion™ Conductivity Probes

Warranty: Out of Box

Specifications

Specifications

Yes
Conductivity Standard
100°C
Soluble
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Conductivity Conditioning Solution

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).

Supplemental information
  • MIXTURE LIST-Contains : edisonite

SDS
Documents

Documents

Product Certifications

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.