Learn More
Thermo Scientific™ Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter
Accurately monitor pH, ion concentration, mV, ORP and temperature on two channels with our pH/ISE benchtop meter for advanced lab analysis.
2100.45 € - 3607.95 €
Specifications
Outputs | USB, RS232 |
---|---|
AC Adapter | Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003) |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
10014414
|
Thermo Scientific™
2115001 |
Orion Dual Star pH/ISE dual channel benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, solutions and stand | Dual Star Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 810001 ROSS Storage Solution; 910104, 910107, 910110 pH 4.01, 7.00, 10.01 pH Buffers |
2633.10 €
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
||||
17187715
|
Thermo Scientific™
2115202 |
Orion Dual Star pH/ISE dual channel benchtop meter chloride & pH kit with Orion chloride ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | Dual Star pH/ISE dual channel meter, 9617BNWP Orion chloride ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 941708 Orion 1000 ppm chloride standard (475 mL), 940011 Orion chloride ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate |
3457.20 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15807904
|
Thermo Scientific™
2115203 |
Orion Dual Star pH/ISE dual channel benchtop meter fluoride & pH kit with Orion fluoride ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | Dual Star pH/ISE dual channel meter, 9609BNWP Orion fluoride ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 040908 Orion 10 ppm fluoride standard premixed with TISAB II (475 mL), 040906 Orion 1 ppm fluoride standard premixed with TISAB II (475 mL), 940909 Orion TISAB II total ionic strength adjustment buffer (3.8 L), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate |
3607.95 €
Each |
Estimated Shipment: 06-12-2024
Log in to see stock. |
||||
12013293
|
Thermo Scientific™
2115101 |
Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra glass-body pH electrode, ATC probe and stand | Dual Star pH/ISE dual channel meter, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate |
2512.50 €
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
||||
10450855
|
Thermo Scientific™
2115102 |
Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra Triode epoxy-body pH/ATC electrode | Dual Star pH/ISE dual channel meter, 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, universal power adapter, literature, computer interface cable and meter test certificate |
2592.90 €
Each |
In Stock
Log in to see stock. |
||||
12003293
|
Thermo Scientific™
2115000 |
Orion Dual Star pH/ISE dual channel benchtop meter with ATC probe and stand | Dual Star pH/ISE dual channel meter, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate |
2100.45 €
Each |
In Stock
Log in to see stock. |
||||
Description
Double your measurement capabilities with the Thermo Scientific™ Orion™ Dual Star™ pH and ISE Benchtop Meter. Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements on two available meter channels, ideal for high throughput pH and ion analysis in the lab. Perform up to 6 point pH and ISE calibrations, log up to 1000 data point sets with time/date stamp and easily transfer to a printer or computer. Use the two channels to measure a pH sample and ISE sample, two pH samples or two ISE samples simultaneously or view each channel separately.
The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter’s two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter’s Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.
- View large, individual measurements on a full screen display or quickly toggle to a split screen display to view two measurements at the same time
- Easy-to-follow onscreen text prompts for calibration and setup menus available in a multi-language user interface with English, Spanish, French, German or Italian options
- EZ Startup menu facilitates quick and easy first-time meter setup
- Numeric keypad and menu-specific function keys allow for quick and efficient meter setup and data entry
- Selectable read modes include Auto-Read, On-Ready, At Timed Intervals, Continuous, Value Change in Measurement and Timed for a variety of measurement collection methods for different analytical methods
- Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
- Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
- Non-volatile meter memory holds up to 1000 data points with time and date stamp
- Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
- Stir samples with ease using the meter-controlled Orion Star stirrer probe
- Meter-attachable electrode stand assists with moving and maintaining electrodes
- IP54-rated housing handles splashes and is wall-mountable to save space
Warranty: 3-year meter replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.Warranty
3-year meter warranty
Specifications
USB, RS232 | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.002 | |
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit | |
Yes, adjustable | |
1 point relative mV (RmV) mode, 1 point ORP mode | |
CE, TUV 3-1, FCC Class A | |
1000 with date and time stamp | |
2 | |
0.0001 to 19,900 in ppm, M, mg/L, % or unit-less | |
-2.000 to 19.999 | |
0.1, 0.01, 0.001 | |
Graphic LCD with Backlight | |
7.000 pH (adjustable), 1.000 ISE (adjustable) | |
English, Spanish, French, Italian and German | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Orion Dual Star pH/ISE/mV/RmV/ORP/Temperature Dual Channel Benchtop Meter | |
±1999.9 mV | |
5 to 85 %, non-condensing | |
0.1°C, 0.1°F | |
41°F to 113°F (ambient) | |
1 point temperature offset calibration | |
3 Year Meter Warranty | |
2 lb. | |
90 to 260 VAC 50/60 Hz |
Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003) | |
±0.2 mV or ±0.05% of reading whichever is greater | |
±0.1°C | |
High/low limit alarm | |
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ | |
1 to 6 | |
Transfer single, range or all data points to printer or computer | |
2 to 6 | |
pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature | |
±1999.9 mV | |
0.0001 minimum, 1 to 3 significant digits (user selectable) | |
-5.0°C to +105.0°C | |
Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) | |
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys | |
Automatic data logging with Auto-Read, On Ready, At Timed Intervals, Value Change In Measurement and | |
10 per channel with password protection | |
Universal AC adapter (included) | |
±1999.9 mV | |
0.1 mV | |
Smart Stability and Smart Averaging | |
5°C to 45°C (ambient) | |
Manual or automatic with ATC temperature probe | |
24 x 17 x 9 cm (9.5 x 6.7 x 3.5 in.) | |
0.9 kg |