Learn More
Thermo Scientific™ Orion™ Filling Solutions for Process Analyzer Reference Electrodes
Enhance the performance and extend the life of ion selective electrodes used in our process analyzers with this filling solution.
195.84 €
Specifications
Type Filling Solution
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15827134
View Documents
Thermo Scientific™
181073
Filling Solution for Orion Process Analyzer 100056/210056 Reference Electrodes, 5 x 60mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Enhance the performance and extend the life of ion selective electrodes used in Orion™ process analyzers with Thermo Scientific™ Orion™ Filling Solutions for Process Analyzer Reference Electrodes.
Warranty: Out of box
Specifications
Specifications
Filling Solution
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Reference Electrode Internal Filling Solution
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cesium chloride, triton x-100
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.