Learn More
Thermo Scientific™ Orion™ Fluoroborate Half-cell
Easily and economically rejuvenate the Thermo Scientific™ Orion™ Fluoroborate Half-cell.
Marca: Thermo Scientific™ 9305BN
Descripción
Thermo Scientific™ OrionFluoroborate Half-cell can be easily and economically rejuvenated.
- Calibration Standards catalog number: Consult instruction manual
- Required ISA catalog number: 930711
- Required Reference : 900200
Especificaciones
BNC | |
Fluoroborate Half-Cell | |
Fluoroborate Half-Cell with BNC Connector |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.