Learn More
Thermo Scientific™ Orion™ Gas-Sensing ISE membranes
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Brand:  Thermo Scientific™ 951214
 View more versions of this product
Product Code. 10554454
  • 134.67 € / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gas-Sensing ISE membrane
High Performance Ammonia Electrode 9512HPBNWP, 9512HP
Gas-Sensing ISE Membrane
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.