Learn More
Thermo Scientific™ Orion™ Ion Selective Electrode Accessories
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
102.51 € - 702.00 €
Specifications
For Use With (Equipment) | Ion Selective Electrode |
---|---|
Type | Ion Selective Electrode Accessories |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10461213
|
Thermo Scientific™
9300BNWP |
Handle for ISE Half Cell Plastic Membrane Electrodes wth BNC waterproof connector |
583.91 €
Each |
In Stock
Log in to see stock. |
|||||
10428722
|
Thermo Scientific™
948201 |
Polishing Strips for Solid State Ion Selective Electrodes |
102.51 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
11747697
|
Thermo Scientific™
9700BNWP |
Handle for Ion+ ISE Combination Plastic Membrane Electrodes with BNC waterproof connector | N/A | ||||||
Description
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Electrodes handles are used with plastic membrane combination or half-cell electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes.Specifications
Ion Selective Electrode | |
Ion Selective Electrode Accessories |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.